Thermo Scientific Chemicals

3'-Nitroacetophenone, 98+%, Thermo Scientific Chemicals

Catalog number: A12155.36
500 g, Each
Thermo Scientific Chemicals

3'-Nitroacetophenone, 98+%, Thermo Scientific Chemicals

Catalog number: A12155.36
500 g, Each
Quantity
Catalog number: A12155.36
also known as A12155-36
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
121-89-1
IUPAC Name
1-(3-nitrophenyl)ethan-1-one
Molecular Formula
C8H7NO3
InChI Key
ARKIFHPFTHVKDT-UHFFFAOYSA-N
SMILES
CC(=O)C1=CC=CC(=C1)[N+]([O-])=O
Appearance (Color)
Pale cream to cream to yellow to pale brown
Assay (GC)
≥98.0%
Identification (FTIR)
Conforms
Form
Crystals or powder or crystalline powder or fused solid
Melting Point (clear melt)
75.0-81.0?C

Description

3'-Nitroacetophenone is used as an intermediate in pharmaceutical industry for making drugs.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3′-Nitroacetophenone is used as an intermediate in pharmaceutical industry for making drugs.

Solubility
Soluble in water 0.75 g/L @ 20°C.

Notes
Store in cool, dry place, in well sealed container. Store away from oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text