4-Methylmorpholine, 99%, Thermo Scientific Chemicals
4-Methylmorpholine, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4-Methylmorpholine, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
100 mL
2500 mL
Catalog number A12158.AP
also known as A12158-AP
Price (USD)/ Each
41.65
Online exclusive
45.90 
Save 4.25 (9%)
-
Add to cart
Quantity:
500 mL
Request bulk or custom format
Price (USD)/ Each
41.65
Online exclusive
45.90 
Save 4.25 (9%)
Add to cart
4-Methylmorpholine, 99%, Thermo Scientific Chemicals
Catalog numberA12158.AP
Price (USD)/ Each
41.65
Online exclusive
45.90 
Save 4.25 (9%)
-
Add to cart
Chemical Identifiers
CAS109-02-4
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Assay (GC)≥98.5%
FormLiquid
Identification (FTIR)Conforms
Refractive Index1.4340-1.4370 @ 20?C
4-Methylmorpholine is used as a solvent for dyes, resins, waxes and pharmaceuticals. It acts as a crosslinker in the preparation of polyurethane foams, elastomers and adhesives. It is used as a precursor to prepare N-methylmorpholine N-oxide and morpholinium cation based ionic liquids. It is utilized as corrosion inhibitors and anti-scaling agents in industries.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Methylmorpholine is used as a solvent for dyes, resins, waxes and pharmaceuticals. It acts as a crosslinker in the preparation of polyurethane foams, elastomers and adhesives. It is used as a precursor to prepare N-methylmorpholine N-oxide and morpholinium cation based ionic liquids. It is utilized as corrosion inhibitors and anti-scaling agents in industries.

Solubility
Miscible with water.

Notes
Incompatible with acids, acid chlorides, carbon dioxide and strong oxidizing agents.
RUO – Research Use Only

General References:

  1. Widely used base in peptide chemistry, milder than triethylamine. Recommended for the avoidance of racemization in the mixed anhydride method of coupling: J. Am. Chem. Soc., 88, 1338 (1966); 89, 5012 (1967). Also preferred in many cases for the formation of active esters or the neutralization of acid salts. For peptide reagents, see Appendix 6.
  2. Pancotti, A.; Parapini, S.; Agli, M. D.; Gambini, L.; Galli, C.; Sangiovanni, E.; Basilico, N.; Bosisio, E.; Taramelli, D.; Romeo, S. Discovery of oxybisbenzoylamides as a new class of antimalarial agents. Med. Chem. Commun. 2015, 6 (6), 1173-1177.
  3. Leng, C.; Kish, J. D.; Roberts, J. E.; Dwebi, I.; Chon, N.; Liu, Y. Temperature-Dependent Henry’s Law Constants of Atmospheric Amines. J. Phys. Chem. A 2015, 199 (33), 8884-8891.