Glycine hydrochloride is an inhibitory neurotransmitter in spinal cord, allosteric regulator of NMDA receptors.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Glycine hydrochloride is an inhibitory neurotransmitter in spinal cord, allosteric regulator of NMDA receptors.
Solubility
Soluble in water (>100 g/L).
Notes
Hygroscopic. Store away from strong oxidizing agents, water/ moisture.
RUO – Research Use Only
General References:
- Michael G. Wohl; M.D.; Nathan Pastor, M.D. Adiposis Dolorosa (Dercum's Disease) Treatment Of The Asthenic Phase With Prostigmine And Aminoacetic Acid. Jama. 1938, 110 (16), 1261-1264.
- George L. Maison, M.D. Failure Of Gelatin Or Amino-Acetic Acid To Increase The Work Ability Of Individual Normal Human Muscles. Tetrahedron Letters. 1940, 115 (17), 1439-1441.