Thermo Scientific Chemicals

Glycine hydrochloride, 98%, Thermo Scientific Chemicals

Catalog number: A12162.36
500 g, Each
Thermo Scientific Chemicals

Glycine hydrochloride, 98%, Thermo Scientific Chemicals

Catalog number: A12162.36
500 g, Each
Quantity
Catalog number: A12162.36
also known as A12162-36
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
6000-43-7
IUPAC Name
hydrogen 2-aminoacetic acid chloride
Molecular Formula
C2H6ClNO2
InChI Key
IVLXQGJVBGMLRR-UHFFFAOYSA-N
SMILES
[H+].[Cl-].NCC(O)=O
Appearance (Color)
White
Assay from Supplier's CofA
≥97.5% (NaOH titration)
Melting Point
178-182?C
Form
Crystalline powder

Description

Glycine hydrochloride is an inhibitory neurotransmitter in spinal cord, allosteric regulator of NMDA receptors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Glycine hydrochloride is an inhibitory neurotransmitter in spinal cord, allosteric regulator of NMDA receptors.

Solubility
Soluble in water (>100 g/L).

Notes
Hygroscopic. Store away from strong oxidizing agents, water/ moisture.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text