Thermo Scientific Chemicals

beta-D-Galactose pentaacetate, 98%, Thermo Scientific Chemicals

Catalog number: A12163.14
25 g, Each
Thermo Scientific Chemicals

beta-D-Galactose pentaacetate, 98%, Thermo Scientific Chemicals

Catalog number: A12163.14
25 g, Each
Quantity
Catalog number: A12163.14
also known as A12163-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
4163-60-4
IUPAC Name
[3,4,5,6-tetrakis(acetyloxy)oxan-2-yl]methyl acetate
Molecular Formula
C16H22O11
InChI Key
LPTITAGPBXDDGR-UHFFFAOYNA-N
SMILES
CC(=O)OCC1OC(OC(C)=O)C(OC(C)=O)C(OC(C)=O)C1OC(C)=O
Optical Rotation
23.5 ± 1.0? (C=4 in chloroform)
Appearance (Color)
White to pale cream
Form
Crystals or powder or crystalline powder
Assay (GC)
≥97.5%
Identification (FTIR)
Conforms

Description

It is used as a pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as a pharmaceutical intermediates.

Solubility
Soluble in methanol (50 mg/ml), chloroform, and water.

Notes
Store at room temperature. Store away from oxidizing agent.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text