It is used as a pharmaceutical intermediates.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is used as a pharmaceutical intermediates.
Solubility
Soluble in methanol (50 mg/ml), chloroform, and water.
Notes
Store at room temperature. Store away from oxidizing agent.
RUO – Research Use Only
J Ohlsson; G Magnusson; Jörgen Ohlsson; Göran Magnusson. Galabiosyl donors; efficient synthesis from 1,2,3,4, 6-penta-O-acetyl-beta-D-galactopyranose. Carbohydrate Research. 2000, 329 (1), 49-55.
Cerag Dilek; Charles W. Manke; Esin Gulari. Phase behavior of β-d galactose pentaacetate-carbon dioxide binary system. Fluid Phase Equilibria. 2006, 239, (2), 172-177.
Regioselective 1-O-deacylation can be effected with ammonia in acetonitrile at ambient temperature: Synthesis, 1121 (1985).