N-(2-Bromoethyl)phthalimide, 97+%, Thermo Scientific Chemicals
N-(2-Bromoethyl)phthalimide, 97+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

N-(2-Bromoethyl)phthalimide, 97+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
500 g
Catalog number A12164.22
also known as A12164-22
Price (USD)/ Each
120.00
-
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (USD)/ Each
120.00
Add to cart
N-(2-Bromoethyl)phthalimide, 97+%, Thermo Scientific Chemicals
Catalog numberA12164.22
Price (USD)/ Each
120.00
-
Add to cart
Chemical Identifiers
CAS574-98-1
IUPAC Name2-(2-bromoethyl)-2,3-dihydro-1H-isoindole-1,3-dione
Molecular FormulaC10H8BrNO2
InChI KeyCHZXTOCAICMPQR-UHFFFAOYSA-N
SMILESBrCCN1C(=O)C2=CC=CC=C2C1=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream
FormCrystals or powder or crystalline powder
Identification (FTIR)Conforms
Melting Point (clear melt)80.0-87.0°C
Assay (GC)≥97.0%
N-(2-Bromoethyl)phthalimide is an intermediate used in organic synthesis. It can react with phenyl magnesium bromide to get 2-(2-bromo-ethyl)-3-hydroxy-3-phenyl-isoindolin-1-one.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-(2-Bromoethyl)phthalimide is an intermediate used in organic synthesis. It can react with phenyl magnesium bromide to get 2-(2-bromo-ethyl)-3-hydroxy-3-phenyl-isoindolin-1-one.

Solubility
Soluble in water (224 mg/L at 25°C).

Notes
Store in cool, dry place in a tightly sealed container. Store away from oxidizing agent.
RUO – Research Use Only

General References:

  1. T. Karthick; V. Balachandranb; S. Perumal; A. Nataraj. Rotational isomers, vibrational assignments, HOMO-LUMO, NLO properties and molecular electrostatic potential surface of N-(2 bromoethyl) phthalimide. Journal of Molecular Structure. 2011, 1005 (1-3), 202-213.
  2. De Kimpe N.; Virag M.; Keppens M.; Scheinmann F.; Stachulski A.V. Cyclocondensation Of N-(2-Bromoethyl)Phthalimide With The Dianon Of Isobutyric Acid To A Functionalized 2,3- Dihydrooxazolo2,3-Aisoindol-5(9Bh)-One De Rivative. J. Chem. Res., Synop. 1995, 6, 252-253.