TrypLE™ Select Enzyme (10X), no phenol red
TrypLE™ Select Enzyme (10X), no phenol red
Actual product may vary
Gibco™

TrypLE™ Select Enzyme (10X), no phenol red

Green features
TrypLE™ Select is an animal origin free, recombinant enzyme for dissociating a wide range of adherent mammalian cells, including CHO,Read more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
A1217702500 mL
A1217701100 mL
A121770320 x 100mL
Catalog number A1217702
Price (USD)
752.00
Each
-
Add to cart
Quantity:
500 mL
Customize this product
Price (USD)
752.00
Each
Add to cart
Ask our AI about this Product
TrypLE™ Select is an animal origin free, recombinant enzyme for dissociating a wide range of adherent mammalian cells, including CHO, HEK 293, A549, primary human keratinocytes, and embryonic stem cells. We offer a variety of TrypLE™ formulations for a range of cell culture applications.

Compared to trypsin and other dissociation reagents, Gibco™ TrypLE™ Select is:

Gentle on cells—maintains cell health for reproducible results
Room-temperature stable—convenient to use
Easy to use—substitutes directly into existing protocols, with no inactivation required
Animal origin free (AOF)—pure, consistent, and effective

This TrypLE™ Select is modified as follows:

With
• EDTA

Without
• Phenol Red

The formulation of TrypLE™ solutions is available, although we keep the enzyme concentration confidential.

Gentle on cells
TrypLE™ Select, like trypsin, cleaves peptide bonds on the C-terminal sides of lysine and arginine. However, TrypLE™ Select's exceptional purity increases specificity due to the action of a single enzyme. This reduces damage caused by cleavage from multiple enzymes in trypsin and other extracts.

Room-temperature stable
TrypLE™ Select is room-temperature stable and ready to use. TrypLE™ Select cell dissociation reagents remain stable for 24 months at room temperature or 2–8°C, making storage and handling easy and convenient.

Easy to use
TrypLE™ Select can be substituted for trypsin in existing protocols. In addition, dilution alone inactivates TrypLE™ Select, avoiding the need for trypsin inhibitors, such as FBS.

Animal Origin Free (AOF)
Unlike porcine trypsin and other enzymes, TrypLE™ Select is free of animal-derived components, and is formulated on dedicated animal origin-free equipment.

Product Intended Use
For Research Use or Further Manufacturing. Not for use in diagnostic procedures. Customers using Gibco™ TrypLE™ Select in a manufacturing process, who have a submission with the FDA, may request a letter of authorization from us to reference our Type II Drug Master File (DMF).

cGMP Manufacturing and Quality System
Gibco™ TrypLE™ Select is manufactured at a cGMP compliant facility located in Grand Island, New York. The facility is registered with the FDA as a medical device manufacturer and is certified to ISO 13485 standards.

Concentrated forms of TrypLE™ can be aseptically diluted to 1x using a balanced salt solution containing 1 mM EDTA.
For Research Use or Further Manufacturing. Not for use in diagnostic procedures.
Specifications
Cell TypeMammalian Cells
ChelatorsEDTA
Concentration10 X
Culture TypeAdherent Cell Culture
Green FeaturesEnergy efficient, Sustainable packaging
Inorganic SaltsNo Calcium, No Magnesium, No Sodium Phosphate
Osmolality270 - 320 mOsm/kg
Product LineGibco™
Purity or Quality GradeCell Culture Grade
Quantity500 mL
RemarksRoom Temperature Stable
Shelf Life24 Months
Shipping ConditionAmbient Temperature
SourceAnimal origin-free
Tested ForIn vitro Bioassay
ClassificationAnimal Origin-free
FormLiquid
Product TypeCell Culture Dissociation Reagent
Without AdditivesNo Phenol Red
pH6.9 to 7.4
Unit SizeEach
Contents & Storage
Storage conditions: 15–30°C. Protect from light.
Alternate storage conditions: 2–8°C or -5 to -20°C.
Shipping conditions: Ambient.
Shelf life: 24 months from date of manufacture.
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Frequently asked questions (FAQs)

Can I use TrypLE Select Enzyme to detach cells from glass surfaces?

Yes, TrypLE Select Enzyme can detach cells from glass surfaces.

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.

What is the concentration of TrypLE Select Enzyme (10X), no phenol red (Cat. No. A1217701, A1217702, A1217703)?

TrypLE Select Enzyme (10X), no phenol red (Cat. No. A1217701, A1217702, A1217703) contains 100 µg/mL of the rProtease enzyme.

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.

How can I assess residual TrypLE reagent remaining after my downstream processing steps?

We offer the Residual TrypLE ELISA Kit (Cat. No. A6533101) that can specifically detect the recombinant protease (rProtease) in TrypLE reagent.

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.

What enzymes do you recommend for passaging mesenchymal stem cells (MSC)?

We recommend using TrypLE Express Enzyme or TrypLE Select Enzyme for passaging mesenchymal stem cells (MSCs). The usage of trypsin will result in lower cell viability upon passage. TrypLE Express and TrypLE Select are defined, animal origin-free, and stable at both room and refrigerated temperatures.

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.

Is there an upper limit for the amount of time cells can be incubated with TrypLE Express Enzyme, TrypLE Select Enzyme, or CTS TrypLE Select Enzyme for dissociation?

There is no upper limit for TrypLE enzyme incubation. You can incubate your cells with TrypLE longer to detach cells. 30-45 min at 37 degrees C will be usually fine. Or you can consider using TrypLE Select Enzyme (10X) (Cat. No. A1217701 or A1217702 or A1217703) or CTS TrypLE Select Enzyme (Cat. No. A4738001) depending upon the application.

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link