Thermo Scientific Chemicals

(Benzoylmethylene)triphenylphosphorane, 98+%, Thermo Scientific Chemicals

Catalog number: A12184.06
5 g, Each
Thermo Scientific Chemicals

(Benzoylmethylene)triphenylphosphorane, 98+%, Thermo Scientific Chemicals

Catalog number: A12184.06
5 g, Each
Quantity
Catalog number: A12184.06
also known as A12184-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
859-65-4
IUPAC Name
1-phenyl-2-(triphenyl-λ⁵-phosphanylidene)ethan-1-one
Molecular Formula
C26H21OP
InChI Key
MZRSAJZDYIISJW-UHFFFAOYSA-N
SMILES
O=C(C=P(C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1)C1=CC=CC=C1
Appearance (Color)
White to cream
Form
Powder
Melting Point (clear melt)
182-187?C
Assay (HPLC)
≥98.0%
Clarity
2.5% w/v solution in Ethanol: Clear to very faintly turbid

Description

It is used in organic synthesis, wittig reagent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used in organic synthesis, wittig reagent.

Solubility
Soluble in Methanol (almost transparency).

Notes
Air sensitive. Store in refrigerator. Store away from oxidizing agent, air.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text