Thermo Scientific Chemicals

4-Iodobenzotrifluoride, 98%, Thermo Scientific Chemicals

Catalog number: A12189.03
1 g, Each
Thermo Scientific Chemicals

4-Iodobenzotrifluoride, 98%, Thermo Scientific Chemicals

Catalog number: A12189.03
1 g, Each
Quantity
Catalog number: A12189.03
also known as A12189-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
455-13-0
IUPAC Name
1-iodo-4-(trifluoromethyl)benzene
Molecular Formula
C7H4F3I
InChI Key
SKGRFPGOGCHDPC-UHFFFAOYSA-N
SMILES
FC(F)(F)C1=CC=C(I)C=C1
Refractive Index
1.5165-1.5205 @ 20?C
Appearance (Color)
Clear colorless to orange
Form
Liquid
Assay (GC)
≥97.5%

Description

4-Iodobenzotrifluoride may be employed as substrate with electron-deficient aromatic ring, during Mizoroki-Heck reaction with acrylic acid, to afford 4-trifluoromethylcinnnamic acid. It undergoes aminocarbonylation in DMF using phosphoryl chloride to give N,N-dimethyl-(4-trifluoromethyl)benzamide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Iodobenzotrifluoride may be employed as substrate with electron-deficient aromatic ring, during Mizoroki-Heck reaction with acrylic acid, to afford 4-trifluoromethylcinnnamic acid. It undergoes aminocarbonylation in DMF using phosphoryl chloride to give N,N-dimethyl-(4-trifluoromethyl)benzamide.

Solubility
Insoluble in water.

Notes
Light sensitive. Incompatible with strong oxidizing agents. Store in dark.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text