Thermo Scientific Chemicals

N-Benzyldimethylamine, 98+%, Thermo Scientific Chemicals

Catalog number: A12191.AK
250 mL, Each
Thermo Scientific Chemicals

N-Benzyldimethylamine, 98+%, Thermo Scientific Chemicals

Catalog number: A12191.AK
250 mL, Each
Quantity
Catalog number: A12191.AK
also known as A12191-AK
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
103-83-3
IUPAC Name
benzyldimethylamine
Molecular Formula
C9H13N
InChI Key
XXBDWLFCJWSEKW-UHFFFAOYSA-N
SMILES
CN(C)CC1=CC=CC=C1
Appearance (Color)
Clear colorless to pale yellow
Form
Liquid
Assay (GC)
≥98.0%
Identification (FTIR)
Conforms
Refractive Index
1.4995-1.5035 @ 20?C

Description

N,N-Dimethylbenzylamine is used in the preparation of bis[(N,N-dimethylamino)benzyl] selenide. It acts as a catalyst in the curing reaction of formulations of diglycidyl ether of bisphenol A and tetrahydrophthalic anhydride. It undergoes directed ortho metalation with butyl lithium. It reacts with methyl iodide to get ammonium salt, which is used as phase transfer catalysts. Further, it is used as a catalyst for the formation of polyurethane foams and epoxy resins.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N,N-Dimethylbenzylamine is used in the preparation of bis[(N,N-dimethylamino)benzyl] selenide. It acts as a catalyst in the curing reaction of formulations of diglycidyl ether of bisphenol A and tetrahydrophthalic anhydride. It undergoes directed ortho metalation with butyl lithium. It reacts with methyl iodide to get ammonium salt, which is used as phase transfer catalysts. Further, it is used as a catalyst for the formation of polyurethane foams and epoxy resins.

Solubility
Slightly miscible with water.

Notes
Air sensitive. Incompatible with strong acids, strong bases, strong oxidizing agents, strong reducing agents and carbon dioxide.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text