N-Benzyldimethylamine, 98+%, Thermo Scientific Chemicals
N-Benzyldimethylamine, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

N-Benzyldimethylamine, 98+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1000 mL
250 mL
5000 mL
Catalog number A12191.AU
also known as A12191-AU
Price (USD)/ Each
104.65
Online exclusive
116.00 
Save 11.35 (10%)
-
Add to cart
Quantity:
1000 mL
Request bulk or custom format
Price (USD)/ Each
104.65
Online exclusive
116.00 
Save 11.35 (10%)
Add to cart
N-Benzyldimethylamine, 98+%, Thermo Scientific Chemicals
Catalog numberA12191.AU
Price (USD)/ Each
104.65
Online exclusive
116.00 
Save 11.35 (10%)
-
Add to cart
Chemical Identifiers
CAS103-83-3
IUPAC Namebenzyldimethylamine
Molecular FormulaC9H13N
InChI KeyXXBDWLFCJWSEKW-UHFFFAOYSA-N
SMILESCN(C)CC1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to pale yellow
FormLiquid
Assay (GC)≥98.0%
Identification (FTIR)Conforms
Refractive Index1.4995-1.5035 @ 20?C
N,N-Dimethylbenzylamine is used in the preparation of bis[(N,N-dimethylamino)benzyl] selenide. It acts as a catalyst in the curing reaction of formulations of diglycidyl ether of bisphenol A and tetrahydrophthalic anhydride. It undergoes directed ortho metalation with butyl lithium. It reacts with methyl iodide to get ammonium salt, which is used as phase transfer catalysts. Further, it is used as a catalyst for the formation of polyurethane foams and epoxy resins.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N,N-Dimethylbenzylamine is used in the preparation of bis[(N,N-dimethylamino)benzyl] selenide. It acts as a catalyst in the curing reaction of formulations of diglycidyl ether of bisphenol A and tetrahydrophthalic anhydride. It undergoes directed ortho metalation with butyl lithium. It reacts with methyl iodide to get ammonium salt, which is used as phase transfer catalysts. Further, it is used as a catalyst for the formation of polyurethane foams and epoxy resins.

Solubility
Slightly miscible with water.

Notes
Air sensitive. Incompatible with strong acids, strong bases, strong oxidizing agents, strong reducing agents and carbon dioxide.
RUO – Research Use Only

General References:

  1. Harisha, A. S.; Nayak, S. P.; Nagarajan, K.; Row, T. N. G.; Hosamani, A. A. Novel triethylamine mediated thermal reactions of 3-aryl-2-cyanoprop-2-enoic acid derivatives-demethylation, reduction and vinylogation. Tetrahedron Lett. 2015, 56 (11), 1427-1431.
  2. Guo, H.; Zheng, J.; Gan, J.; Liang, L.; Wu, K.; Lu, M. Relationship between crosslinking structure and low dielectric constant of hydrophobic epoxies based on substituted biphenyl mesogenic units. RSC Adv. 2015, 5 (107), 88014-88020.