Thermo Scientific Chemicals

1H-Indene, 90+%, stab. with 0.01% 4-tert-butylcatechol, Thermo Scientific Chemicals

Catalog number: A12192.AD
50 mL, Each
Thermo Scientific Chemicals

1H-Indene, 90+%, stab. with 0.01% 4-tert-butylcatechol, Thermo Scientific Chemicals

Catalog number: A12192.AD
50 mL, Each
Quantity
Catalog number: A12192.AD
also known as A12192-AD
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
95-13-6
IUPAC Name
1H-indene
Molecular Formula
C9H8
InChI Key
YBYIRNPNPLQARY-UHFFFAOYSA-N
SMILES
C1C=CC2=CC=CC=C12
Assay (GC)
≥90.0%
Appearance (Color)
Clear colorless to yellow to brown
Comment
May contain some polymer
Refractive Index
1.5660-1.5760 @ 20?C
Form
Liquid

Description

1H-Indene is a building block that has been used in the synthesis of isatins. It was used in the synthesis of new C60 derivative, indene-C60 bisadduct. It was used in preparing polyindene by the controlled cationic polymerization initiated with cumyl methyl ether/TiCl4 in CH2Cl2.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1H-Indene is a building block that has been used in the synthesis of isatins. It was used in the synthesis of new C60 derivative, indene-C60 bisadduct. It was used in preparing polyindene by the controlled cationic polymerization initiated with cumyl methyl ether/TiCl4 in CH2Cl2.

Solubility
Soluble in ethanol, acetone, ethyl ether, and ether. Insoluble in water.

Notes
Store at 4°C. Store in dark. Incompatible with oxidizing agent, light, heat.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text