Trifluoroacetic acid, 99%, Thermo Scientific Chemicals
Trifluoroacetic acid, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Trifluoroacetic acid, 99%, Thermo Scientific Chemicals

Solvent, catalyst, strong non-oxidizing acid
Have Questions?
Change viewbuttonViewtableView
Quantity:
2500 g
100 g
500 g
Catalog number A12198.0E
also known as A12198-0E
Price (USD)/ Each
602.65
Online exclusive
669.00 
Save 66.35 (10%)
-
Add to cart
Quantity:
2500 g
Request bulk or custom format
Price (USD)/ Each
602.65
Online exclusive
669.00 
Save 66.35 (10%)
Add to cart
Trifluoroacetic acid, 99%, Thermo Scientific Chemicals
Catalog numberA12198.0E
Price (USD)/ Each
602.65
Online exclusive
669.00 
Save 66.35 (10%)
-
Add to cart
Chemical Identifiers
CAS76-05-1
IUPAC Nametrifluoroacetic acid
Molecular FormulaC2HF3O2
InChI KeyDTQVDTLACAAQTR-UHFFFAOYSA-N
SMILESOC(=O)C(F)(F)F
View more
SpecificationsSpecification SheetSpecification Sheet
Assay from Supplier's CofA≥98.5% (US-sourced material)
Water Content (Karl Fischer Titration)≤0.5%
Appearance (Color)Clear colorless to pale yellow
Assay (Aqueous acid-base Titration)≥98.5 to ≤101.5% (UK-sourced material)
CommentMaterial sourced in UK and US
View more
The combination of properties like solubility in most of the solvents, volatility, catalytic property, and strong acidity with non-oxidizing nature makes it a widely used reagent in organic synthesis. Trifluoroacetic acid is an important building block in the synthesis of pharmaceuticals, agrochemicals and performance products. It is a precursor to many fluorinated compounds, and widely used in peptide synthesis and other organic transformations involving deprotection of t-BOC group. TFA finds use as an ion pairing agent in liquid chromatography, as a solvent in NMR spectroscopy and as a calibrant in mass spectrometry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
The combination of properties like solubility in most of the solvents, volatility, catalytic property, and strong acidity with non-oxidizing nature makes it a widely used reagent in organic synthesis. Trifluoroacetic acid is an important building block in the synthesis of pharmaceuticals, agrochemicals and performance products. It is a precursor to many fluorinated compounds, and widely used in peptide synthesis and other organic transformations involving deprotection of t-BOC group. TFA finds use as an ion pairing agent in liquid chromatography, as a solvent in NMR spectroscopy and as a calibrant in mass spectrometry.

Solubility
Miscible with ether, acetone, ethanol, benzene, hexane, and CCl4

Notes
Trifluoroacetic acid (TFA) is a stable, hygroscopic, volatile and low-boiling organic acid. Storage in a cool place is recommended.
RUO – Research Use Only

General References:

  1. López, S. E.; Salazar, J. Trifluoroacetic acid: Uses and recent applications in organic synthesis. J. Fluorine Chem. 2013, 156, 73-100.
  2. Natu, A. D.; Burde, A. S.; Limaye, R. A.; Paradkar, M. V. Acceleration of the Dakin reaction by trifluoroacetic acid. J. Chem. Res. 2014, 38 (6), 325-386.