Thermo Scientific Chemicals

1,3-Indanedione, 97%, Thermo Scientific Chemicals

Catalog number: A12206.14
25 g, Each
Thermo Scientific Chemicals

1,3-Indanedione, 97%, Thermo Scientific Chemicals

Catalog number: A12206.14
25 g, Each
Quantity
Catalog number: A12206.14
also known as A12206-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
606-23-5
IUPAC Name
2,3-dihydro-1H-indene-1,3-dione
Molecular Formula
C9H6O2
InChI Key
UHKAJLSKXBADFT-UHFFFAOYSA-N
SMILES
O=C1CC(=O)C2=CC=CC=C12
Appearance (Color)
White to cream or yellow to yellow-green to pale green to pale brown
Form
Powder or crystals
Assay (HPLC)
≥96.0%
Identification (FTIR)
Conforms
Melting Point (clear melt)
124-131?C

Description

It is used in the first stage of forensic identification of latent fingerprints. It is particularly useful for paper, and for items printed with thermal inks such as receipts.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used in the first stage of forensic identification of latent fingerprints. It is particularly useful for paper, and for items printed with thermal inks such as receipts.

Solubility
Soluble in ethanol, ether, benzene, slightly soluble in water.

Notes
Store in cool, dry place in a tightly sealed container. Store away from oxidizing agent.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text