Thermo Scientific Chemicals

2,6-Dimethoxybenzaldehyde, 98+%, Thermo Scientific Chemicals

Catalog number: A12209.14
25 g, Each
Thermo Scientific Chemicals

2,6-Dimethoxybenzaldehyde, 98+%, Thermo Scientific Chemicals

Catalog number: A12209.14
25 g, Each
Quantity
Catalog number: A12209.14
also known as A12209-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
3392-97-0
IUPAC Name
2,6-dimethoxybenzaldehyde
Molecular Formula
C9H10O3
InChI Key
WXSGQHKHUYTJNB-UHFFFAOYSA-N
SMILES
COC1=CC=CC(OC)=C1C=O
Appearance (Color)
Pale cream to cream to yellow
Form
Crystals or powder or crystalline powder
Assay (GC)
≥98.0%
Free acid (titration)
≤1%
Melting Point (clear melt)
92.0-98.0?C

Description

2,6-Dimethoxybenzaldehyde has been used in the preparation of 2,6-dihydroxybenzaldehyde by demethylation with AlBr3.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,6-Dimethoxybenzaldehyde has been used in the preparation of 2,6-dihydroxybenzaldehyde by demethylation with AlBr3.

Solubility
Soluble in Methanol (almost transparency).

Notes
Air sensitive. Store in cool, dry place in a tightly sealed container. Store away from oxidizing agent, air.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text