3'-(Trifluoromethyl)acetophenone, 98+%, Thermo Scientific Chemicals
3'-(Trifluoromethyl)acetophenone, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

3'-(Trifluoromethyl)acetophenone, 98+%, Thermo Scientific Chemicals

Have Questions?
Quantity:
5 g
Catalog number A12210.06
also known as A12210-06
Price (USD)/ Each
51.65
Online exclusive
57.10 
Save 5.45 (10%)
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
51.65
Online exclusive
57.10 
Save 5.45 (10%)
Add to cart
3'-(Trifluoromethyl)acetophenone, 98+%, Thermo Scientific Chemicals
Catalog numberA12210.06
Price (USD)/ Each
51.65
Online exclusive
57.10 
Save 5.45 (10%)
-
Add to cart
Chemical Identifiers
CAS349-76-8
IUPAC Name1-[3-(trifluoromethyl)phenyl]ethan-1-one
Molecular FormulaC9H7F3O
InChI KeyABXGMGUHGLQMAW-UHFFFAOYSA-N
SMILESCC(=O)C1=CC=CC(=C1)C(F)(F)F
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to pale yellow
FormLiquid
Assay (GC)≥98.0%
Refractive Index1.4615-1.4655 @ 20?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only