The Amplex Red Monoamine Oxidase Assay Kit provides a sensitive and simple fluorometric method for detecting monoamine oxidase activity in purified enzyme preparations at levels as low as 12 μU/mL and in tissue samples with as little as 20 μg protein in a 100 μL assay volume.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Detection Method
Fluorescent
For Use With (Application)
Monoamine Oxidase Assay
For Use With (Equipment)
Fluorometer, Microplate Reader
Product Type
Red Monoamine Oxidase Assay Kit
Dye Type
Other Label(s) or Dye(s)
Format
Cuvettes, 96-well plate
Product Line
Amplex™
Shipping Condition
Room Temperature
Contents & Storage
Store in freezer -5°C to -30°C and protect from light.