Thermo Scientific Chemicals

2-Furonitrile, 98+%, Thermo Scientific Chemicals

Catalog number: A12214.14
25 g, Each
Thermo Scientific Chemicals

2-Furonitrile, 98+%, Thermo Scientific Chemicals

Catalog number: A12214.14
25 g, Each
Quantity
Catalog number: A12214.14
also known as A12214-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
617-90-3
IUPAC Name
furan-2-carbonitrile
Molecular Formula
C5H3NO
InChI Key
YXDXXGXWFJCXEB-UHFFFAOYSA-N
SMILES
N#CC1=CC=CO1
Form
Liquid
Refractive Index
1.4775-1.4815 @ 20°C
Appearance (Color)
Clear colorless to pale yellow
Assay (GC)
≥98.0%

Description

2-Furonitrile is used as a extractive distillation solvent and as a sweetening agent. It is also used as an intermediate in pharmaceutical and fine chemical synthesis. 2-Furonitrile was employed as substrate to investigate the substrate specificity of nitrilase from Rhodococcus rhodochrous Jl cell.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Furonitrile is used as a extractive distillation solvent and as a sweetening agent. It is also used as an intermediate in pharmaceutical and fine chemical synthesis. 2-Furonitrile was employed as substrate to investigate the substrate specificity of nitrilase from Rhodococcus rhodochrous Jl cell.

Solubility
Soluble in most common organic solvents. Slightly soluble in water.

Notes
Store in cool, dry place in a tightly sealed container. Store away from oxidizing agent.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text