Invitrogen™

Amplex™ Acetylcholine/Acetlycholinesterase Assay Kit

Catalog number: A12217
Have Questions?
Invitrogen™

Amplex™ Acetylcholine/Acetlycholinesterase Assay Kit

Catalog number: A12217
Have Questions?
Catalog Number
A12217
Unit Size
500 assays
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
A12217500 assays
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
The Amplex™ Red Acetylcholine/Acetylcholinesterase Assay Kit provides an ultrasensitive method for detecting acetylcholinesterase (AChE) activity in a fluorescence microplate reader or fluorometer.

See our complete line of Fluorescence Microplate assays.

• Detect AChE activity levels as low as 0.002 U/mL in one hour
• Detect acetylcholine levels as low as 0.3 μM using excess AChE
• Achieve detection ranges of 0.3 μM to 100 μM acetylcholine
• Format allows for multiple time point measurements
• Designed for minimal autofluorescence interference

AChE activity is monitored indirectly using 10-acetyl-3,7-dihydroxyphenoxazine (Amplex™ Red reagent), a sensitive fluorogenic probe for hydrogen peroxide. AChE converts the acetylcholine substrate to choline, which is then oxidized by choline oxidase to betaine and hydrogen peroxide. In the presence of horseradish peroxidase, hydrogen peroxide reacts with the Amplex™ Red reagent in a 1:1 stoichiometric ratio to generate the highly fluorescent product resorufin.

Because resorufin has absorption and fluorescence emission maxima of approximately 571 nm and 585 nm, respectively, there is little interference from autofluorescence in most biological samples. Experiments with purified AChE from electric eel indicate that the Amplex™ Red Acetylcholine/Acetylcholinesterase Assay Kit can detect AChE levels as low as 0.002 U/mL using a reaction time of one hour. By providing an excess of AChE in the assay, the kit can also be used to detect acetylcholine levels as low as 0.3 μM, with a range of detection from 0.3 μM to 100 μM acetylcholine.

Use Amplex™ Red Assays for a Broad Range of Investigations
A wide variety of validated Amplex™ Red assays are available for studying cell signaling and lipids, neurobiology, inflammation and immune function, and metabolism. We also offer Amplex™ UltraRed Reagent (Cat. No. A36006), a second-generation reagent providing greater sensitivity and brighter fluorescence, and the Amplex™ Red/UltraRed Stop Reagent (Cat. No. A33855). The Amplex™ Red/UltraRed Stop Reagent provides convenience and control by allowing the fluorescence signal-generating reaction to be terminated at a user-determined time point. After addition of the stop reagent, the fluorescence signal remains stable for at least three hours. Custom assay design and packaging are also available.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Detection Method
Fluorescence Intensity
For Use With (Application)
Acetylcholine/Acetlycholinesterase Assay
For Use With (Equipment)
Fluorescence Microplate Reader
Product Type
Acetylcholine⁄Acetlycholinesterase Assay
Product Line
Amplex™
Shipping Condition
Room Temperature
Substrate Properties
Molecular Substrate
Substrate Type
HRP (Horseradish Peroxidase) Substrate, Acetylcholinesterase Substrate
Target Enzyme
Acetylcholinesterase

Contents & Storage

Store in freezer -5°C to -30°C and protect from light.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text