Thermo Scientific Chemicals

5-Bromo-2-chlorobenzoic acid, 98+%, Thermo Scientific Chemicals

Catalog number: A12218.09
10 g, Each
Thermo Scientific Chemicals

5-Bromo-2-chlorobenzoic acid, 98+%, Thermo Scientific Chemicals

Catalog number: A12218.09
10 g, Each
Quantity
Catalog number: A12218.09
also known as A12218-09
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
21739-92-4
IUPAC Name
5-bromo-2-chlorobenzoate
Molecular Formula
C7H3BrClO2
InChI Key
FGERXQWKKIVFQG-UHFFFAOYSA-M
SMILES
[O-]C(=O)C1=CC(Br)=CC=C1Cl
Melting Point (clear melt)
155.0-162.0?C
Form
Crystals or powder or crystalline powder
Assay (Aqueous acid-base Titration)
≥98.0 to ≤102.0%
Assay (Silylated GC)
≥98.0%
Appearance (Color)
White

Description

5-Bromo-2-chlorobenzoic acid was used to evaluate the homogeneity of organic powder. It can be used to produce 2-chloro-5-deuteriobenzoic acid.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-Bromo-2-chlorobenzoic acid was used to evaluate the homogeneity of organic powder. It can be used to produce 2-chloro-5-deuteriobenzoic acid.

Solubility
Soluble in water 2.63 g/L @20°C.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text