Thermo Scientific Chemicals

4'-Bromoacetophenone, 98%, Thermo Scientific Chemicals

Catalog number: A12222.36
500 g, Each
Thermo Scientific Chemicals

4'-Bromoacetophenone, 98%, Thermo Scientific Chemicals

Catalog number: A12222.36
500 g, Each
Quantity
Catalog number: A12222.36
also known as A12222-36
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
99-90-1
IUPAC Name
1-(4-bromophenyl)ethan-1-one
Molecular Formula
C8H7BrO
InChI Key
WYECURVXVYPVAT-UHFFFAOYSA-N
SMILES
CC(=O)C1=CC=C(Br)C=C1
Form
Crystals or powder or crystalline powder
Assay (GC)
≥97.5%
Identification (FTIR)
Conforms
Melting Point
48.0-54.0?C
Appearance (Color)
White to cream or pale yellow

Description

Labeled 4’-Bromoacetophenone possess activity against positive phototaxis of Chlamydomonas cells. A fundamental starting material for organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Labeled 4’-Bromoacetophenone possess activity against positive phototaxis of Chlamydomonas cells. A fundamental starting material for organic synthesis.

Solubility
Soluble in Chloroform. Insoluble in water.

Notes
Store in cool, dry place in a tightly sealed container. Store away from oxidizing agent, bases.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text