Thermo Scientific Chemicals

1-Dodecanol, 98%, Thermo Scientific Chemicals

Catalog number: A12228.30
250 g, Each
Thermo Scientific Chemicals

1-Dodecanol, 98%, Thermo Scientific Chemicals

Catalog number: A12228.30
250 g, Each
Quantity
Catalog number: A12228.30
also known as A12228-30
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
112-53-8
IUPAC Name
dodecan-1-ol
Molecular Formula
C12H26O
InChI Key
LQZZUXJYWNFBMV-UHFFFAOYSA-N
SMILES
CCCCCCCCCCCCO
Form
Liquid
Appearance (Color)
Clear colorless to white
Assay (GC)
≥97.5%
Identification (FTIR)
Conforms
Refractive Index
1.4400-1.4440 @ 20?C

Description

1-Dodecanol is used as a cosmetic, textile auxiliaries, synthetic oil, emulsifiers and flotation agent of raw materials, a detergent raw material, a foaming agent of the toothpaste.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Dodecanol is used as a cosmetic, textile auxiliaries, synthetic oil, emulsifiers and flotation agent of raw materials, a detergent raw material, a foaming agent of the toothpaste.

Solubility
Soluble in water (0.004g/L).

Notes
Store in cool, dry place in a tightly sealed container. Store away from oxidizing agent.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text