Thermo Scientific Chemicals

Succinic anhydride, 99%, Thermo Scientific Chemicals

Catalog number: A12245.0I
5000 g, Each
Thermo Scientific Chemicals

Succinic anhydride, 99%, Thermo Scientific Chemicals

Catalog number: A12245.0I
5000 g, Each
Quantity
Catalog number: A12245.0I
also known as A12245-0I
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
108-30-5
IUPAC Name
oxolane-2,5-dione
Molecular Formula
C4H4O3
InChI Key
RINCXYDBBGOEEQ-UHFFFAOYSA-N
SMILES
O=C1CCC(=O)O1
Identification (FTIR)
Conforms
Melting Point (clear melt)
116-121?C
Form
Powder or crystals or crystalline powder or flakes and/or chips
Assay (Aqueous acid-base Titration)
≥98.5 to ≤101.5%
Appearance (Color)
White

Description

Succinic anhydride is used as a sizing agent and curative for epoxy resin. It acts as an additive in paper production. It is utilized in the preparation of covalently cross-linked oxidized-alginate and N-succinyl-chitosan hydrogels. It serves as an intermediate for functionalized oxide surfaces on a chip and for polyester resins. Further, it finds use in cosmetics, pharmaceuticals and in coatings.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Succinic anhydride is used as a sizing agent and curative for epoxy resin. It acts as an additive in paper production. It is utilized in the preparation of covalently cross-linked oxidized-alginate and N-succinyl-chitosan hydrogels. It serves as an intermediate for functionalized oxide surfaces on a chip and for polyester resins. Further, it finds use in cosmetics, pharmaceuticals and in coatings.

Solubility
Soluble in chloroform.Soluble in chloroform, carbon tetrachloride and alcohol. Slightly soluble in ether. Insoluble in water.

Notes
Moisture sensitive. Incompatible with strong oxidizing agents, alcohols and amines.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text