Thermo Scientific Chemicals

2-Chloro-N-(hydroxymethyl)acetamide, 98%, Thermo Scientific Chemicals

Catalog number: A12247.18
50 g, Each
Thermo Scientific Chemicals

2-Chloro-N-(hydroxymethyl)acetamide, 98%, Thermo Scientific Chemicals

Catalog number: A12247.18
50 g, Each
Quantity
Catalog number: A12247.18
also known as A12247-18
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
2832-19-1
Appearance (Color)
White to cream
Form
Crystals or powder or crystalline powder
Assay (HPLC)
≥97.5%
Melting Point (clear melt)
99.0-105.0?C

Description

An organic building block. It is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals. As dyestuff intermediate .

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
An organic building block. It is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals. As dyestuff intermediate .

Solubility
Very soluble in water. Poorly soluble in aliphatic hydrocarbons.

Notes
Hygroscopic. Store in cool, dry conditions in well sealed containers. Store away from strong oxidizing agents. Keep container tightly closed.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text