Thermo Scientific Chemicals

4-Chlorobutyryl chloride, 98%, Thermo Scientific Chemicals

Catalog number: A12249.0B
1000 g, Each
Thermo Scientific Chemicals

4-Chlorobutyryl chloride, 98%, Thermo Scientific Chemicals

Catalog number: A12249.0B
1000 g, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: A12249.0B
also known as A12249-0B
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
4635-59-0
IUPAC Name
4-chlorobutanoyl chloride
Molecular Formula
C4H6Cl2O
InChI Key
CDIIZULDSLKBKV-UHFFFAOYSA-N
SMILES
ClCCCC(Cl)=O
Appearance (Color)
Clear colorless to yellow
Form
Liquid
Assay (GC)
≥97.5%
Refractive Index
1.4595-1.4635 @ 20?C

Description

4-Chlorobutyryl chloride (4-CBCl) is mainly used in the production of pharmaceuticals and agrochemicals. It is used as pyrazinamide intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Chlorobutyryl chloride (4-CBCl) is mainly used in the production of pharmaceuticals and agrochemicals. It is used as pyrazinamide intermediate.

Solubility
soluble in diethyl ether, ethanol, slightly soluble in water.

Notes
Moisture sensitive. Store under inert gas. Store away from oxidizing agent, bases,
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text