Thermo Scientific Chemicals

Salicylic acid, 99%, Thermo Scientific Chemicals

Catalog number: A12253.36
500 g, Each
Thermo Scientific Chemicals

Salicylic acid, 99%, Thermo Scientific Chemicals

Catalog number: A12253.36
500 g, Each
Quantity
Catalog number: A12253.36
also known as A12253-36
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
69-72-7
IUPAC Name
2-hydroxybenzoic acid
Molecular Formula
C7H6O3
InChI Key
YGSDEFSMJLZEOE-UHFFFAOYSA-N
SMILES
OC(=O)C1=CC=CC=C1O
Appearance (Color)
White to cream
Form
Crystals or powder or crystalline powder
Melting Point (clear melt)
156-163?C
Assay (Aqueous acid-base Titration)
≥98.5 to ≤101.5%
Assay (Silylated GC)
≥98.5%

Description

Salicylic acid is used as a food preservative as well as in the treatment of many skin disorders such as acne, dandruff, psoriasis and seborrheic dermatitis of the skin and scalp. It is an antiseptic. It is mainly used in the pharmaceutical industry for the manufacture of antipyretic, analgesic, anti-inflammatory and diuretic drugs. It is also used in dye industry for making azo direct dye and for acidic mordant dyeing.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Salicylic acid is used as a food preservative as well as in the treatment of many skin disorders such as acne, dandruff, psoriasis and seborrheic dermatitis of the skin and scalp. It is an antiseptic. It is mainly used in the pharmaceutical industry for the manufacture of antipyretic, analgesic, anti-inflammatory and diuretic drugs. It is also used in dye industry for making azo direct dye and for acidic mordant dyeing.

Solubility
Soluble in carbon terachloride, benzene, propanol, absolute ethanol, acetone, toluene and ether. Slightly soluble in water.

Notes
Light sensitive. Incompatible with strong oxidizing agents, strong bases, iodine, iron and iron salts.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text