3,4-Dichlorophenol, 97%, Thermo Scientific Chemicals
3,4-Dichlorophenol, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

3,4-Dichlorophenol, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number A12256.14
also known as A12256-14
Price (USD)/ Each
76.70
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
76.70
Add to cart
3,4-Dichlorophenol, 97%, Thermo Scientific Chemicals
Catalog numberA12256.14
Price (USD)/ Each
76.70
-
Add to cart
Chemical Identifiers
CAS95-77-2
IUPAC Name3,4-dichlorophenol
Molecular FormulaC6H4Cl2O
InChI KeyWDNBURPWRNALGP-UHFFFAOYSA-N
SMILESOC1=CC=C(Cl)C(Cl)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream to brown
Assay (GC)≥96.0%
Water Content (Karl Fischer Titration)≤0.75%
FormCrystals or powder or crystalline powder or flakes or needles or fibres
Melting Point (clear melt)62.0-69.0°C
It is also a photo-degradation product of the common antibacterial and antifungal agent triclosan along with the dioxin 2,8-dichlorodibenzo-p-dioxin.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is also a photo-degradation product of the common antibacterial and antifungal agent triclosan along with the dioxin 2,8-dichlorodibenzo-p-dioxin.

Solubility
Slightly soluble ion water 9.2 g/L at 20degC. Soluble in aqueous alkali, oxygenated and chlorinated solvents. Soluble in ethanol, benzene, chloroform and ethyl ether.

Notes
Store in cool, dry conditions in well sealed containers. Store away from strong oxidizing agents, acid, acid fumes, acid chlorides, acid anhydrides, caustics. Keep container tightly closed. Store in an area without drain or sewer access. Separated from strong oxidants and food and feedstuffs.
RUO – Research Use Only

General References:

  1. K Valli,; M H Gold. Degradation of 2,4-dichlorophenol by the lignin-degrading fungus Phanerochaete chrysosporium. J. Bacteriol. 1991, 173 (1) ,345-352 .
  2. W. Z. Tang,; C. P. Huang. 2,4-Dichlorophenol Oxidation Kinetics by Fenton's Reagent. Environmental Technology. 1996, 17 (12), 1371-1378.