1-Bromo-3-ethylbenzene, 98%, Thermo Scientific Chemicals
1-Bromo-3-ethylbenzene, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

1-Bromo-3-ethylbenzene, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
1 g
5 g
Catalog number A12258.14
also known as A12258-14
Price (USD)/ Each
636.00
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
636.00
Add to cart
1-Bromo-3-ethylbenzene, 98%, Thermo Scientific Chemicals
Catalog numberA12258.14
Price (USD)/ Each
636.00
-
Add to cart
Chemical Identifiers
CAS2725-82-8
IUPAC Name1-bromo-3-ethylbenzene
Molecular FormulaC8H9Br
InChI KeyZRFJYAZQMFCUIX-UHFFFAOYSA-N
SMILESCCC1=CC=CC(Br)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
FormLiquid
Assay (GC)≥97.5%
Refractive Index1.5430-1.5470 @ 20?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only