Thermo Scientific Chemicals

2-Bromo-5-methoxybenzoic acid, 98+%, Thermo Scientific Chemicals

Catalog number: A12261.30
250 g, Each
Thermo Scientific Chemicals

2-Bromo-5-methoxybenzoic acid, 98+%, Thermo Scientific Chemicals

Catalog number: A12261.30
250 g, Each
Quantity
Catalog number: A12261.30
also known as A12261-30
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
22921-68-2
IUPAC Name
2-bromo-5-methoxybenzoate
Molecular Formula
C8H6BrO3
InChI Key
ODHJOROUCITYNF-UHFFFAOYSA-M
SMILES
COC1=CC=C(Br)C(=C1)C([O-])=O
Assay (Silylated GC)
>98.0%
Assay (Aqueous acid-base Titration)
>98.0 to <102.0%
Appearance (Color)
White to cream
Form
Crystalline powder

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text