Thermo Scientific Chemicals

Pyridine-2,6-dicarboxylic acid, 98%, Thermo Scientific Chemicals

Catalog number: A12263.14
25 g, Each
Thermo Scientific Chemicals

Pyridine-2,6-dicarboxylic acid, 98%, Thermo Scientific Chemicals

Catalog number: A12263.14
25 g, Each
Quantity
Catalog number: A12263.14
also known as A12263-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
499-83-2
IUPAC Name
pyridine-2,6-dicarboxylic acid
Molecular Formula
C7H5NO4
InChI Key
WJJMNDUMQPNECX-UHFFFAOYSA-N
SMILES
OC(=O)C1=CC=CC(=N1)C(O)=O
Appearance (Color)
White to pale cream
Form
Crystals or powder or crystalline powder
Assay (Aqueous acid-base Titration)
≥97.5 to ≤102.5% (Non-U.S. specification)
Assay (Silylated GC)
≥97.5% (Non-U.S. Specification)
Assay from Suppliers CofA
≥97.5% (HPLC, U.S. specification)

Description

Pyridine-2,6-dicarboxylic acid is used in the preparation of dipicolinato ligated lanthanide and transition metal complexes. It acts as a chelating agent for chromium, zinc, manganese, copper, iron and molybdenum. Its calcium-dipcolinic acid complex is used to protect deoxyribonucleic acid (DNA) from heat denaturation which enhances the DNA stability. It plays an important role as a marker for the effectiveness of sterilization.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Pyridine-2,6-dicarboxylic acid is used in the preparation of dipicolinato ligated lanthanide and transition metal complexes. It acts as a chelating agent for chromium, zinc, manganese, copper, iron and molybdenum. Its calcium-dipcolinic acid complex is used to protect deoxyribonucleic acid (DNA) from heat denaturation which enhances the DNA stability. It plays an important role as a marker for the effectiveness of sterilization.

Solubility
Insoluble in water.

Notes
Incompatible with acids, bases and oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text