Thermo Scientific Chemicals

delta-Valerolactam, 98+%, Thermo Scientific Chemicals

Catalog number: A12271.14
25 g, Each
Thermo Scientific Chemicals

delta-Valerolactam, 98+%, Thermo Scientific Chemicals

Catalog number: A12271.14
25 g, Each
Quantity
Catalog number: A12271.14
also known as A12271-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
675-20-7
IUPAC Name
piperidin-2-one
Molecular Formula
C5H9NO
InChI Key
XUWHAWMETYGRKB-UHFFFAOYSA-N
SMILES
O=C1CCCCN1
Form
Crystalline or Fused solid
Assay (GC)
>97.5%
Solution Test
Clear (5% w/v in Ethanol)
Water Content (Karl Fischer Titration)
<1.0%
Appearance (Color)
White to yellow or brown

Description

delta-Valerolactam is the starting material for a new route to hexahydrojulolidines, using intramolecular 1,4-cycloaddition. It is used as an intermediate in the preparation of other chemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
delta-Valerolactam is the starting material for a new route to hexahydrojulolidines, using intramolecular 1,4-cycloaddition. It is used as an intermediate in the preparation of other chemicals.

Solubility
Soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text