Thermo Scientific Chemicals

Trisodium citrate dihydrate, 99%, Thermo Scientific Chemicals

Catalog number: A12274.0E
2500 g, Each
Thermo Scientific Chemicals

Trisodium citrate dihydrate, 99%, Thermo Scientific Chemicals

Catalog number: A12274.0E
2500 g, Each
Quantity
Catalog number: A12274.0E
also known as A12274-0E
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
6132-04-3
IUPAC Name
trisodium 2-hydroxypropane-1,2,3-tricarboxylate dihydrate
Molecular Formula
C6H9Na3O9
InChI Key
NLJMYIDDQXHKNR-UHFFFAOYSA-K
SMILES
O.O.[Na+].[Na+].[Na+].OC(CC([O-])=O)(CC([O-])=O)C([O-])=O
Form
Crystalline powder or crystals
Assay from Supplier's CofA
≥98.5% (U.S. sourced material)
Identification (FTIR)
Conforms (non-U.S. sourced material)
Assay (Non-aqueous acid-base Titration)
≥98.5 to ≤101.5% (non-U.S. sourced material)
Comment
Material sourced in the U.S. and in other countries

Description

Trisodium citrate dihydrate, is widely applied in food, beverages and fillers as a buffering, sequestering or an emulsifying agent. It used as an anticoagulant in blood transfusions, osmotic laxative, functional fluids, solvents cleaning, furnishing care products, laundry dishwashing products and cleaning automobile radiators.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Trisodium citrate dihydrate, is widely applied in food, beverages and fillers as a buffering, sequestering or an emulsifying agent. It used as an anticoagulant in blood transfusions, osmotic laxative, functional fluids, solvents cleaning, furnishing care products, laundry dishwashing products and cleaning automobile radiators.

Solubility
Soluble in water. Insoluble in alcohol.

Notes
Stable and incompatible with bases, reducing agents, oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text