Description
2-Bromomesitylene is a useful synthetic intermediate, Aromatics, Boron Derivatives, Catalyst. It is an intermediate used to prepare Hydrogen[4-[bis(2,4,6-trimethylphenyl)phosphino]-2,3,5,6-tetrafluorophenyl]hydrobis(2,3,4,5,6-pentafluorophenyl)borate (H714510) which is a frustrated phosphonium borate for metal-free catalytic hydrogenation of imines.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2-Bromomesitylene is a useful synthetic intermediate, Aromatics, Boron Derivatives, Catalyst. It is an intermediate used to prepare Hydrogen[4-[bis(2,4,6-trimethylphenyl)phosphino]-2,3,5,6-tetrafluorophenyl]hydrobis(2,3,4,5,6-pentafluorophenyl)borate (H714510) which is a frustrated phosphonium borate for metal-free catalytic hydrogenation of imines.
Solubility
Insoluble in water(20°C) .
Notes
Light Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with Strong oxidizing agents, excess heat.
RUO – Research Use Only