Thermo Scientific Chemicals

2-Bromomesitylene, 99%, Thermo Scientific Chemicals

Catalog number: A12277.22
100 g, Each
Thermo Scientific Chemicals

2-Bromomesitylene, 99%, Thermo Scientific Chemicals

Catalog number: A12277.22
100 g, Each
Quantity
Catalog number: A12277.22
also known as A12277-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
576-83-0
IUPAC Name
2-bromo-1,3,5-trimethylbenzene
Molecular Formula
C9H11Br
InChI Key
RRTLQRYOJOSPEA-UHFFFAOYSA-N
SMILES
CC1=CC(C)=C(Br)C(C)=C1
Refractive Index
1.5505-1.5535 @ 20?C
Appearance (Color)
Clear colorless to pale yellow
Assay (GC)
≥98.5%
Identification (FTIR)
Conforms
Form
Liquid

Description

2-Bromomesitylene is a useful synthetic intermediate, Aromatics, Boron Derivatives, Catalyst. It is an intermediate used to prepare Hydrogen[4-[bis(2,4,6-trimethylphenyl)phosphino]-2,3,5,6-tetrafluorophenyl]hydrobis(2,3,4,5,6-pentafluorophenyl)borate (H714510) which is a frustrated phosphonium borate for metal-free catalytic hydrogenation of imines.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Bromomesitylene is a useful synthetic intermediate, Aromatics, Boron Derivatives, Catalyst. It is an intermediate used to prepare Hydrogen[4-[bis(2,4,6-trimethylphenyl)phosphino]-2,3,5,6-tetrafluorophenyl]hydrobis(2,3,4,5,6-pentafluorophenyl)borate (H714510) which is a frustrated phosphonium borate for metal-free catalytic hydrogenation of imines.

Solubility
Insoluble in water(20°C) .

Notes
Light Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with Strong oxidizing agents, excess heat.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text