Dibenzothiophene, 98%, Thermo Scientific Chemicals
Dibenzothiophene, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Dibenzothiophene, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Catalog number A12288.14
also known as A12288-14
Price (USD)/ Each
48.65
Online exclusive
53.90 
Save 5.25 (10%)
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
48.65
Online exclusive
53.90 
Save 5.25 (10%)
Add to cart
Dibenzothiophene, 98%, Thermo Scientific Chemicals
Catalog numberA12288.14
Price (USD)/ Each
48.65
Online exclusive
53.90 
Save 5.25 (10%)
-
Add to cart
Chemical Identifiers
CAS132-65-0
IUPAC Name8-thiatricyclo[7.4.0.0²,⁷]trideca-1(9),2(7),3,5,10,12-hexaene
Molecular FormulaC12H8S
InChI KeyIYYZUPMFVPLQIF-UHFFFAOYSA-N
SMILESS1C2=C(C=CC=C2)C2=C1C=CC=C2
View more
SpecificationsSpecification SheetSpecification Sheet
FormPowder
Appearance (Color)White to pale cream or pale yellow
Identification (FTIR)Conforms
Solution Test5% in acetone; clear to slightly hazy yellow solution
Assay (GC)≥97.5%
Dibenzothiophene is used to investigate the effect of sulfur compounds in gasoline range during the fluid catalytic cracking (FCC) process.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Dibenzothiophene is used to investigate the effect of sulfur compounds in gasoline range during the fluid catalytic cracking (FCC) process.

Solubility
Soluble in water.

Notes
Incompatible with strong oxidizing agents.
RUO – Research Use Only

General References:

  1. Lithiation occurs at the 4-position (ortho to the S atom). The lithio-derivative reacts with electrophiles, such as bromine: J. Am. Chem. Soc., 76, 5786 (1954).
  2. Trehoux, A.; Roux, Y.; Guillot, R.; Mahy, J. P.; Avenier, F. Catalytic oxidation of dibenzothiophene and thioanisole by a diiron(III) complex and hydrogen peroxide. J. Mol. Catal. A: Chem. 2015, 396, 40-46.
  3. Fan, C.; Duan, C.; Wei, Y.; Ding, D.; Xu, H.; Huang, W. Dibenzothiophene-Based Phosphine Oxide Host and Electron-Transporting Materials for Efficient Blue Thermally Activated Delayed Fluorescence Diodes through Compatibility Optimization. Chem. Mater. 2015, 27 (14), 5131-5140.