Thermo Scientific Chemicals

Dibenzothiophene, 98%, Thermo Scientific Chemicals

Catalog number: A12288.36
500 g, Each
Thermo Scientific Chemicals

Dibenzothiophene, 98%, Thermo Scientific Chemicals

Catalog number: A12288.36
500 g, Each
Quantity
Catalog number: A12288.36
also known as A12288-36
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
132-65-0
IUPAC Name
8-thiatricyclo[7.4.0.0²,⁷]trideca-1(9),2(7),3,5,10,12-hexaene
Molecular Formula
C12H8S
InChI Key
IYYZUPMFVPLQIF-UHFFFAOYSA-N
SMILES
S1C2=C(C=CC=C2)C2=C1C=CC=C2
Form
Powder
Appearance (Color)
White to pale cream or pale yellow
Identification (FTIR)
Conforms
Solution Test
5% in acetone; clear to slightly hazy yellow solution
Assay (GC)
≥97.5%

Description

Dibenzothiophene is used to investigate the effect of sulfur compounds in gasoline range during the fluid catalytic cracking (FCC) process.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Dibenzothiophene is used to investigate the effect of sulfur compounds in gasoline range during the fluid catalytic cracking (FCC) process.

Solubility
Soluble in water.

Notes
Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text