Thermo Scientific Chemicals

Potassium oxalate hydrate, 99%, Thermo Scientific Chemicals

Catalog number: A12289.0B
1000 g, Each
Thermo Scientific Chemicals

Potassium oxalate hydrate, 99%, Thermo Scientific Chemicals

Catalog number: A12289.0B
1000 g, Each
Quantity
Catalog number: A12289.0B
also known as A12289-0B
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
6487-48-5
IUPAC Name
dipotassium hydrate oxalate
Molecular Formula
C2H2K2O5
InChI Key
QCPTVXCMROGZOL-UHFFFAOYSA-L
SMILES
O.[K+].[K+].[O-]C(=O)C([O-])=O
Appearance (Color)
White
Assay (Non-aqueous acid-base Titration)
>98.5 to <102.5% (dry wt. basis)
Assay (Redox Titration)
>98.5 to <102.5% (dry wt. basis)
Loss on Drying
6.7 - 12.6% (200?C/constant wt., monohydrate 9.78%)
Form
Crystals or crystalline powder

Description

Potassium oxalate hydrate is used as a chelating agent, a purifying agent in the pharmaceutical industry and a precipitating agent in rare-earth metal processing. It finds application as a grinding agent, as a rust remover in metal treatment and as a bleaching agent in the textile and wood industry. It is also used in waste water treatment and as a scale remover in automobile radiators. Furthermore, it is used as a reagent in analytical chemistry for the determination of calcium and other heavy metals as well as reducing agents in photography. It is also involved in the preparation of SBA-15 (nanostructured SiO2) supported vanadium oxide catalyst.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Potassium oxalate hydrate is used as a chelating agent, a purifying agent in the pharmaceutical industry and a precipitating agent in rare-earth metal processing. It finds application as a grinding agent, as a rust remover in metal treatment and as a bleaching agent in the textile and wood industry. It is also used in waste water treatment and as a scale remover in automobile radiators. Furthermore, it is used as a reagent in analytical chemistry for the determination of calcium and other heavy metals as well as reducing agents in photography. It is also involved in the preparation of SBA-15 (nanostructured SiO2) supported vanadium oxide catalyst.

Solubility
Soluble in water.

Notes
Hygroscopic. Incompatible with halogens, ammonia, cyanides and heavy metals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text