Thermo Scientific Chemicals

1-Bromo-2,4-dichlorobenzene, 98%, Thermo Scientific Chemicals

Catalog number: A12298.06
5 g, Each
Thermo Scientific Chemicals

1-Bromo-2,4-dichlorobenzene, 98%, Thermo Scientific Chemicals

Catalog number: A12298.06
5 g, Each
Quantity
Catalog number: A12298.06
also known as A12298-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1193-72-2
IUPAC Name
1-bromo-2,4-dichlorobenzene
Molecular Formula
C6H3BrCl2
InChI Key
ISHYFWKKWKXXPL-UHFFFAOYSA-N
SMILES
ClC1=CC=C(Br)C(Cl)=C1
Appearance (Color)
Clear colorless to white to pale yellow
Assay (GC)
≥97.5%
Form
Liquid
Refractive Index
1.5965-1.6005 @ 20?C

Description

1-Bromo-2,4-dichlorobenzene is used to produce 2,4,4'-trichloro-biphenyl. This reaction will need aq. Na2CO3 and solvents dioxane, ethanol. It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Bromo-2,4-dichlorobenzene is used to produce 2,4,4′-trichloro-biphenyl. This reaction will need aq. Na2CO3 and solvents dioxane, ethanol. It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields.

Solubility
Slightly soluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text