Thermo Scientific Chemicals

(Dichloromethylene)dimethylammonium chloride, 95%, Thermo Scientific Chemicals

Catalog number: A12300.14
25 g, Each
Thermo Scientific Chemicals

(Dichloromethylene)dimethylammonium chloride, 95%, Thermo Scientific Chemicals

Catalog number: A12300.14
25 g, Each
Quantity
Catalog number: A12300.14
also known as A12300-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
33842-02-3
IUPAC Name
dichloro(dimethylamino)methylium chloride
Molecular Formula
C3H6Cl3N
InChI Key
NRNFKRFWZQQDMD-UHFFFAOYSA-M
SMILES
[Cl-].CN(C)[C+](Cl)Cl
Appearance (Color)
White to pale yellow
Form
Crystals or powder or crystalline powder
Assay (Titration ex Chloride)
≥94.0 to ≤106.0%

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text