Ethyl 3-methyl-1H-pyrazole-5-carboxylate, 97%, Thermo Scientific Chemicals
Ethyl 3-methyl-1H-pyrazole-5-carboxylate, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Ethyl 3-methyl-1H-pyrazole-5-carboxylate, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number A12304.06
also known as A12304-06
Price (USD)/ Each
147.00
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
147.00
Add to cart
Ethyl 3-methyl-1H-pyrazole-5-carboxylate, 97%, Thermo Scientific Chemicals
Catalog numberA12304.06
Price (USD)/ Each
147.00
-
Add to cart
Chemical Identifiers
CAS4027-57-0
IUPAC Nameethyl 5-methyl-1H-pyrazole-3-carboxylate
Molecular FormulaC7H10N2O2
InChI KeyBOTXQJAHRCGJEG-UHFFFAOYSA-N
SMILESCCOC(=O)C1=NNC(C)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale yellow or pale cream
Assay (GC)≥96.0%
FormCrystals or powder or crystalline powder
Melting Point (clear melt)77.0-87.0°C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only