Thermo Scientific Chemicals

Methyl 2,4,6-trihydroxybenzoate, 98%, Thermo Scientific Chemicals

Catalog number: A12305.22
100 g, Each
Thermo Scientific Chemicals

Methyl 2,4,6-trihydroxybenzoate, 98%, Thermo Scientific Chemicals

Catalog number: A12305.22
100 g, Each
Quantity
Catalog number: A12305.22
also known as A12305-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
3147-39-5
IUPAC Name
methyl 2,4,6-trihydroxybenzoate
Molecular Formula
C8H8O5
InChI Key
AQDIJIAUYXOCGX-UHFFFAOYSA-N
SMILES
COC(=O)C1=C(O)C=C(O)C=C1O
Assay (Silylated GC)
≥97.5%
Appearance (Color)
White to cream
Form
Powder

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text