o-Cresol is mainly used as a precursor to other compounds. Kolbe-Schmitt carboxylation gives o-cresotinic acid, a pharmaceutical intermediate. The muscle relaxant Mephenesin is an ether derived from o-cresol. Used as adhesives and sealant chemicals, fuels and fuel additives, intermediates, Solvents, Paints and coatings.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
o-Cresol is mainly used as a precursor to other compounds. Kolbe-Schmitt carboxylation gives o-cresotinic acid, a pharmaceutical intermediate. The muscle relaxant Mephenesin is an ether derived from o-cresol. Used as adhesives and sealant chemicals, fuels and fuel additives, intermediates, Solvents, Paints and coatings.
Solubility
Soluble in chloroform, ether. Insoluble in water but soluble in about 40 parts water. Soluble in carbon tetrachloride
Notes
Store in cool, dry conditions in well sealed containers. Store away from strong oxidizing agents. Keep container tightly closed.
RUO – Research Use Only
General References:
- Boatto G, Nieddu M,; Carta A, Pau A,; Lorenzoni S,; Manconi P,; Serra D. Determination of phenol and o-cresol by GC/MS in a fatal poisoning case.. Forensic Sci Int. year of publication , 139 (2-3), 191-194.
- Hua-Wei Chen,; Young Ku,; Agus Irawan. Photodecomposition of o-cresol by UV-LED/TiO2 process with controlled periodic illumination. Chemosphere. 2007, 69 (2), 184-190.