Thermo Scientific Chemicals

2-Bromotoluene, 99%, Thermo Scientific Chemicals

Catalog number: A12315.22
100 g, Each
Thermo Scientific Chemicals

2-Bromotoluene, 99%, Thermo Scientific Chemicals

Catalog number: A12315.22
100 g, Each
Quantity
Catalog number: A12315.22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
95-46-5
IUPAC Name
1-bromo-2-methylbenzene
Molecular Formula
C7H7Br
InChI Key
QSSXJPIWXQTSIX-UHFFFAOYSA-N
SMILES
CC1=CC=CC=C1Br
Form
Liquid
Refractive Index
1.5550-1.5580 @ 20?C
Appearance (Color)
Clear colorless
Assay (GC)
≥98.5%
Identification (FTIR)
Conforms

Description

2-Bromotoluene was used in the synthesis of (±)-isocomene. Suzuki coupling of 2-bromotoluene with phenylboronic acid using palladium nanoparticles supported on alumina-based oxides as catalyst has been reported. 2-Bromotoluene is used in manufacturing pharmaceuticals (especially for nonsteroidal antiinflammatory drugs; Meclofenamic acid, Mefenamic acid) and other organic compounds

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Bromotoluene was used in the synthesis of (±)-isocomene. Suzuki coupling of 2-bromotoluene with phenylboronic acid using palladium nanoparticles supported on alumina-based oxides as catalyst has been reported. 2-Bromotoluene is used in manufacturing pharmaceuticals (especially for nonsteroidal antiinflammatory drugs; Meclofenamic acid, Mefenamic acid) and other organic compounds

Solubility
Soluble in alcohol, ether, acetone, and ethanol. Insoluble in water.

Notes
Hygroscopic. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents. Store at 4°C.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text