4-Bromo-3-methylanisole, 97%, Thermo Scientific Chemicals
4-Bromo-3-methylanisole, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4-Bromo-3-methylanisole, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
10 g
250 g
Catalog number A12318.18
also known as A12318-18
Price (USD)/ Each
204.00
-
Add to cart
Quantity:
50 g
Request bulk or custom format
Price (USD)/ Each
204.00
Add to cart
4-Bromo-3-methylanisole, 97%, Thermo Scientific Chemicals
Catalog numberA12318.18
Price (USD)/ Each
204.00
-
Add to cart
Chemical Identifiers
CAS27060-75-9
IUPAC Name1-bromo-4-methoxy-2-methylbenzene
Molecular FormulaC8H9BrO
InChI KeyBLZNSXFQRKVFRP-UHFFFAOYSA-N
SMILESCOC1=CC=C(Br)C(C)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to pale yellow
FormLiquid
Refractive Index1.5580-1.5630 @ 20°C
Assay (GC)≥96.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only