Thermo Scientific Chemicals

2,6-Dichloroaniline, 98%, Thermo Scientific Chemicals

Catalog number: A12320.36
500 g, Each
Thermo Scientific Chemicals

2,6-Dichloroaniline, 98%, Thermo Scientific Chemicals

Catalog number: A12320.36
500 g, Each
Quantity
Catalog number: A12320.36
also known as A12320-36
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
608-31-1
IUPAC Name
2,6-dichloroaniline
Molecular Formula
C6H5Cl2N
InChI Key
JDMFXJULNGEPOI-UHFFFAOYSA-N
SMILES
NC1=C(Cl)C=CC=C1Cl
Form
Crystals or powder or crystalline powder or chunks or fused solid
Assay (GC)
≥97.5%
Melting Point (clear melt)
35.0-42.0?C
Appearance (Color)
White to cream to pale gray

Description

2,6-Dichloroaniline is used in the pharmaceutical industry for the synthesis of antibacterial agents of lomefloxacin, Lee uric acid and clonidine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,6-Dichloroaniline is used in the pharmaceutical industry for the synthesis of antibacterial agents of lomefloxacin, Lee uric acid and clonidine.

Solubility
Soluble in water (1.6 g/L at 20°C), dimethyl sulfoxide and methanol.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text