Thermo Scientific Chemicals

Pyridoxal-5-phosphate hydrate, 98%, Thermo Scientific Chemicals

Catalog number: A12323.03
1 g, Each
Thermo Scientific Chemicals

Pyridoxal-5-phosphate hydrate, 98%, Thermo Scientific Chemicals

Catalog number: A12323.03
1 g, Each
Quantity
Catalog number: A12323.03
also known as A12323-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
41468-25-1
IUPAC Name
[(4-formyl-5-hydroxy-6-methylpyridin-3-yl)methoxy]phosphonic acid
Molecular Formula
C8H10NO6P
InChI Key
NGVDGCNFYWLIFO-UHFFFAOYSA-N
SMILES
CC1=NC=C(COP(O)(O)=O)C(C=O)=C1O
Identification (FTIR)
Conforms (UK-sourced material)
Appearance (Color)
Pale cream to cream to pale yellow
Assay (HPLC)
≥97.5%
Form
Powder or fused solid
Water Content (Karl Fischer Titration)
≤15.41% (UK-sourced material)

Description

Active form of Vitamin B-6Pyridoxal-5’-phosphate monohydrate is used to modify the lysyl and valyl residues in proteins. It acts as a coenzyme in transamination reactions and as an inhibitor for purinergic receptors and intracellular. It is involved in decarboxylation and deamination reactions viz. the conversion of dopa into dopamine of glutamate into GABA and histidine to histamine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Active form of Vitamin B-6Pyridoxal-5’-phosphate monohydrate is used to modify the lysyl and valyl residues in proteins. It acts as a coenzyme in transamination reactions and as an inhibitor for purinergic receptors and intracellular. It is involved in decarboxylation and deamination reactions viz. the conversion of dopa into dopamine of glutamate into GABA and histidine to histamine.

Solubility
Soluble in alkaline solutions. Sparingly soluble in water.

Notes
Light sensitive. Store in cool place. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong acids.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text