Thermo Scientific Chemicals

1-Bromo-3,5-bis(trifluoromethyl)benzene, 98%, Thermo Scientific Chemicals

Catalog number: A12337.18
50 g, Each
Thermo Scientific Chemicals

1-Bromo-3,5-bis(trifluoromethyl)benzene, 98%, Thermo Scientific Chemicals

Catalog number: A12337.18
50 g, Each
Quantity
Catalog number: A12337.18
also known as A12337-18
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
328-70-1
IUPAC Name
1-bromo-3,5-bis(trifluoromethyl)benzene
Molecular Formula
C8H3BrF6
InChI Key
CSVCVIHEBDJTCJ-UHFFFAOYSA-N
SMILES
FC(F)(F)C1=CC(=CC(Br)=C1)C(F)(F)F
Assay (GC)
≥97.5%
Identification (FTIR)
Conforms
Appearance (Color)
Clear colorless
Refractive Index
1.4255-1.4295 @ 20?C
Form
Liquid

Description

1,3-Bis(trifluoromethyl)-5-bromobenzene is used to prepare the tetrakis[3,5-bis(trifluoromethyl)phenyl]borate ion, which acts as a stabilizing counterion for electrophilic organic and organometallic cations.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,3-Bis(trifluoromethyl)-5-bromobenzene is used to prepare the tetrakis[3,5-bis(trifluoromethyl)phenyl]borate ion, which acts as a stabilizing counterion for electrophilic organic and organometallic cations.

Solubility
Immiscible with water.

Notes
Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text