Thermo Scientific Chemicals

Zirconium dichloride oxide octahydrate, 98%, Thermo Scientific Chemicals

Catalog number: A12342.0E
2500 g, Each
Thermo Scientific Chemicals

Zirconium dichloride oxide octahydrate, 98%, Thermo Scientific Chemicals

Catalog number: A12342.0E
2500 g, Each
Quantity
Catalog number: A12342.0E
Price (USD)
Have Questions?

Chemical Identifiers

CAS
13520-92-8
Molecular Formula
Cl2H18O9Zr
InChI Key
FUBACIUATZGHAC-UHFFFAOYSA-N
SMILES
O.O.O.O.O.O.O.O.Cl.Cl.O=[Zr]
Molecular Weight (g/mol)
324.26
Appearance (Color)
White to pale yellow
Form
Crystals or powder or crystalline powder or fused solid
Assay (Titration ex Chloride)
≥97.5 to ≤102.5%
Identification (FTIR)
Conforms
Water Content (Karl Fischer Titration)
37.76-50.28% (6-10 waters)

Description

Zirconium dichloride oxide is utilized as a precursor to prepare other zirconium compounds. It is employed in acid dyes, pigment toners and antiperspirants. It finds application as a rubber additive and a fiber treatment agent. It is also used in paint drying, refractories, ceramics and glaze.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Zirconium dichloride oxide is utilized as a precursor to prepare other zirconium compounds. It is employed in acid dyes, pigment toners and antiperspirants. It finds application as a rubber additive and a fiber treatment agent. It is also used in paint drying, refractories, ceramics and glaze.

Solubility
Soluble in water and alcohol.

Notes
Hygroscopic, moisture sensitive and incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text