Zirconium dichloride oxide octahydrate, 98%, Thermo Scientific Chemicals
Zirconium dichloride oxide octahydrate, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Zirconium dichloride oxide octahydrate, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
2500 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number A12342.36
Price (USD)/ Each
-
Quantity:
500 g
Request bulk or custom format
Chemical Identifiers
CAS13520-92-8
Molecular FormulaCl2H18O9Zr
InChI KeyFUBACIUATZGHAC-UHFFFAOYSA-N
SMILESO.O.O.O.O.O.O.O.Cl.Cl.O=[Zr]
Molecular Weight (g/mol)324.26
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale yellow
FormCrystals or powder or crystalline powder or fused solid
Assay (Titration ex Chloride)≥97.5 to ≤102.5%
Identification (FTIR)Conforms
Water Content (Karl Fischer Titration)37.76-50.28% (6-10 waters)
Zirconium dichloride oxide is utilized as a precursor to prepare other zirconium compounds. It is employed in acid dyes, pigment toners and antiperspirants. It finds application as a rubber additive and a fiber treatment agent. It is also used in paint drying, refractories, ceramics and glaze.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Zirconium dichloride oxide is utilized as a precursor to prepare other zirconium compounds. It is employed in acid dyes, pigment toners and antiperspirants. It finds application as a rubber additive and a fiber treatment agent. It is also used in paint drying, refractories, ceramics and glaze.

Solubility
Soluble in water and alcohol.

Notes
Hygroscopic, moisture sensitive and incompatible with strong oxidizing agents.
RUO – Research Use Only

General References:

  1. Hu, Q.; Hu, W.; Kong, J.; Zhang, X. Ultrasensitive electrochemical DNA biosensor by exploiting hematin as efficient biomimetic catalyst toward in situmetallization. Biosens. Bioelectron. 2015, 63, 269-275.
  2. Yan, X.; Lu, N.; Fan, B.; Bao, J.; Pan, D.; Wang, M.; Li, R. Synthesis of mesoporous and tetragonal zirconia with inherited morphology from metal-organic frameworks. CrystEngComm. 2015, 17 (33), 6426-6433.